Learn More
Sepax Technologies Inc Sepax HPLC Column SRT-C SEC-1000 PEEK, 5 µm, 4.6 x 150 mm
SDP
Supplier:  Sepax Technologies Inc 235950P4615
Encompass
SRT-C SEC phase, developed based on innovative surface coating technology, comprises of uniform, hydrophilic, and neutral nanometer thick film chemically bonded to high purity and mechanically stabilized silica. SRT-C, uses lay-down monolayer on porous silica as oppose to SRT of stand-up monolayer, offers ideal phase chemistry for hydrophobic samples like insulin, membrane protein monoclonal antibodies derivatized with polymer branches (polypeptide or PEG) separation.
Catalog No. 50-233-7412