Learn More
Sepax Technologies Inc Sepax HPLC Column Zenix SEC-150, 3 µm, 7.8 x 200 mm
SDP
Supplier:  Sepax Technologies Inc 2131507820
Encompass
Made of 3 µm silica particles, Zenix SEC packing is a breakthrough technology for size exclusion chromatography. Combination of 3 µm silica particle size and proprietary surface technology enables highest separation efficiency and resolution for biological molecules and water soluble polymers. Available pore sizes of Zenix packings are 80, 100, 150 and 300 Å
Catalog No. 50-233-6833