Learn More
Medchemexpress LLC HY-138638 25g , Sephadex LH 20 CAS:9041-37-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13863825G
Encompass_Preferred
HY-138638 25g Sephadex LH 20 CAS: 9041-37-6 Sephadex LH 20 could be used for the isolation of natural compounds and food, such as red wine and pigments. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1430