Learn More
Sigma Aldrich Fine Chemicals Biosciences SepharoseR 4B Cytiva 17 01
Supplier:  Sigma Aldrich Fine Chemicals Biosciences GE17012001
Encompass
Sepharose(R) 4B is a well-proven Agarose gel filtration base matrix and is frequently used for coupling affinity ligands to the matrix. The matrix is not pre-activated and the user performs all steps in coupling.
Catalog No. 50-290-7627