Learn More
Medchemexpress LLC Sepimostat (FUT-187 free base) | 103926-64-3 | 98.0% | C21H19N5O2 | 10MG
SDP
Supplier:  Medchemexpress LLC HY13629910MG
Encompass_Preferred
Sepimostat is a small-molecule NR2B-selective NMDA receptor antagonist reported to exhibit neuroprotective activity by antagonizing the Ifenprodil-binding site. It is supplied as an off-white to yellow solid with reported purity suitable for research and analytical applications.
  • Acts as an NR2B-selective NMDA receptor antagonist.
  • Exhibits neuroprotective activity in reported studies.
  • Inhibits Ifenprodil binding with a reported Ki of 27.7 μM.
  • Off-white to yellow solid formulation suitable for analytical use.
  • High purity for reproducible experimental results.
  • Available in formats suitable for small-scale research workflows.
Catalog No. 50-002-59934