Learn More
Microsolv Technology Corporation Septa, 20mm, Headspace, White Silicone Rubber / Beige PTFE. For use in 20mm Crimp Caps, sold separately. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502507
Encompass_Preferred
Silicone Rubber / PTFE Septa with Durometer of 45 Shore A, (hardness) and 3.0mm thick with Ultra Clean PTFE which is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Use in Aluminum or Steel Crimp Caps for Headspace autosamplers. Supplied in Polyethylene Bags.
Catalog No. 50-226-9898