Learn More
Trajan Scientific Septa Inlet Marathon Long Life 24/PK
SDP
Supplier:  Trajan Scientific 041914
Encompass_Preferred
12 7 mm (1/2 ) OD Marathon long-life center guide GC septa PK24
Catalog No. 50-004-94181