Learn More
Chemglass Life Sciences 8-425 Replacement Septa, Minum-Ware
SDP
Supplier:  Chemglass Life Sciences MW10411
Encompass_Preferred
8-425 Replacement Septa, Minum-Ware. Replacement septa for use with MW-102 open-top screw caps on Minum-Ware glassware components. Septa are made of silicone rubber with one side having a RED PTFE face, while the opposite side has an ultrathin covering of polypropylene to prevent needle coring when penetrating septa.
Catalog No. 50-153-2674