Learn More
Perkin Elmer US LLC 13 mm Silicone Septa for Screw Top Waste and Wash Vial, 100/pk
Supplier:  Perkin Elmer US LLC N9302780
Encompass_Preferred
Vials and closures are a critical part of your workflow; just as important as the instrument or column used. It is easy to think of a vial or cap as an inexpensive commodity that won’t impact your results, but the reality is different. It’s not only the physical attributes that can affect overall analytical performance, but also the quality of the glass and septa used. Contamination from sub-standard glass or poor quality septa (figure 3) can lead to interferences, inaccuracies and failures which ultimately effects lab productivity.
Catalog No. 50-905-1907