Learn More
Apexbio Technology LLC [Ser25] Protein Kinase C (19-31) 136795-05-6 25mg
SDP
Supplier:  Apexbio Technology LLC A103325
Encompass_Preferred
[Ser25] Protein Kinase C (19-31) (CAS 136795-05-6) is an oligopeptide derived from the pseudosubstrate regulatory domain (amino acids 19-31) of Protein Kinase C alpha (PKC ) modified by substitution of alanine with serine at residue 25 Its sequence is Arg-Phe-Ala-Arg-Lys-Gly-Ser-Leu-Arg-Gln-Lys-Asn-Val with a molecular weight of 1559 82 and molecular formula C67H118N26O17 It acts as a PKC substrate peptide enabling in vitro measurement of kinase enzymatic activity via serine phosphorylation PKC enzymes participate in diverse cellular processes including adhesion transformation cell cycle control and calcium signaling making this peptide an important tool for kinase activity studies and intracellular signaling research
Catalog No. 50-000-63685