Learn More
Selleck Chemical LLC Serabelisib (TAK-117) 5mg 1268454-23-4 INK-1117, MLN-1117
SDP
Supplier:  Selleck Chemical LLC S85815MG
Encompass_Preferred
Serabelisib (TAK-117, INK-1117, MLN-1117) is a potent and selective oral PI3K? isoform inhibitor (IC50 of 21 nmol/L against PI3K?) that has demonstrated > 100-fold selectivity relative to other class I PI3K family members (PI3K?/?/?) and mTOR, and a high degree of selectivity against many other kinase. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8214