Learn More
Medchemexpress LLC Serabelisib (MLN1117) | 1268454-23-4 | 99.4% | 50 MG

Supplier: Medchemexpress LLC HY1228550MG
Serabelisib (MLN1117) is a selective PI3Kα (p110α) inhibitor supplied as a high-purity solid for biochemical and cell-based research applications.
- Selective PI3Kα inhibitor with reported p110α IC50 of 15 nM.
- High purity (99.4%).
- Molecular formula C19H17N5O3 and molecular weight 363.37.
- Supplied in small-scale solid quantities suitable for assay development and preclinical studies.
- Recommended storage: powder at -20 °C (long term) or 4 °C (short term); in solvent at -80 °C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.