Learn More
Cayman Chemical Seralutinib 10MG
SDP
Supplier:  Cayman Chemical 3775410 MG
Encompass_Preferred
A PDGFR, c-Kit, and CSF1R inhibitor; inhibits PDGF-BB-induced phosphorylation of ERK in H1703 cells, HPASMCs, and HLFs (IC50s = 74, 49, and 62 nM, respectively); inhibits stem cell factor-induced c-Kit autophosphorylation in HPAECs and M-CSF-induced CSF1R phosphorylation in human primary differentiated macrophages (IC50s = 7.8 and 14.4 nM, respectively); inhibits the proliferation of H1703 cells and PDGF-BB-induced HPASMCs and HLFs (IC50s = 32, 33, and 29 nM, respectively); prevents increases in pulmonary artery systolic pressure in a rat model of pulmonary arterial hypertension induced by monocrotaline pneumonectomy when administered via passive inhalation at 2.5 mg/kg
Catalog No. 50-291-8114