Learn More
Medchemexpress LLC Seralutinib | 1619931-27-9 | 99.94% | 469.53 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY10919010MM/1ML
Encompass_Preferred
Seralutinib (GB002) is an inhaled PDGFRα and PDGFRβ inhibitor. It also targets CSF1R and c-KIT with IC50s of 8 nM and 14 nM, respectively. This product is used in the study for pulmonary arterial hypertension.
  • Inhaled PDGFRα and PDGFRβ inhibitor
  • Targets CSF1R and c-KIT
  • Used in the study for pulmonary arterial hypertension
  • Significantly reduces right ventricular systolic pressure and mean pulmonary artery pressure
  • Reduces pulmonary arteriole muscularization
  • Restores BMPR2 protein expression in the lung lobes
  • Well tolerated
  • Appearance: Solid, white to light yellow
Catalog No. 50-003-46032