Learn More
Seracare Life Sciences Inc Seraseq ctDNA Mutation Mix V4, 93 Clinically Relevant Variants, 0.1% Variant Allele Frequency, 25 µL
Supplier:  Seracare Life Sciences Inc 07103097
Encompass
The Seraseq ctDNA Mutation Mix v4 AF0.1% is a ready-to-use reference material enhancing NGS assay validation. It contains 93 clinically relevant variants across 71 genes, ensuring accurate detection of various mutations with low background noise. The advanced fragmentation process mimics natural ctDNA fragment sizes, improving confidence in results. Ideal for research and diagnostics, it offers comprehensive monitoring from preparation to analysis, setting it apart for its reliability and precision.
Catalog No. NC3314052