Learn More
Seracare Life Sciences Inc Seraseq FFPE TMB RM Score 13 | 2 x 10 um
Supplier:  Seracare Life Sciences Inc 07101618
Encompass
To improve and harmonize the measurement of TMB, Seracare Life Sciences has generated and characterized tumor-normal human cell lines with different TMB scores for whole exome sequencing analysis of TMB in patient samples, for validating targeted NGS panel-based TMB assays, and as controls for TMB measurements in I-O clinical trial projects.
Catalog No. NC1894280