Learn More
Crescent Chemical Co Inc SERDOLIT MB ANALYTICAL GRDE
SDP
Supplier:  Crescent Chemical Co Inc SE45500.03
Encompass_Preferred
SERDOLIT MB analytical grade Mixed-bed ion exchanger with exhaustion indicatorcomposed of SERDOLIT Blue (OH--form) and SERDOLIT CS-2 (H+-form) in a ratio at approx. 1.5:1 (v/v); ready-to-use, suitable for demineralizing water. Size - 500G Storage
Catalog No. 50-249-104