Learn More
Cayman Chemical SerglIflozIn A 10mg
SDP
Supplier:  Cayman Chemical 1434210 MG
Encompass_Preferred
An SGLT2 inhibitor (Ki = 2.39 nM for the human transporter) and active metabolite of sergliflozin etabonate; selective for SGLT2 over SGLT1 (Ki = 708 nM for the human transporter); co-infusion with glucose reduces maximal renal glucose reabsorption rates in rats
Catalog No. 50-295-5003