Learn More
Alkali Scientific Alkali Scientific™ Serial Printer for Alkali Scientific Series Dx and Tx balances
SDP
Supplier:  Alkali Scientific W3130
Encompass_Preferred
The W3130 Thermal Printer offers a small footprint, modern design, and is compatible with the Alkali Scientific Dx and Tx Analytical Balances.  Connection is easy with the included cables, and weight values and other critical info can be printed quickly and clearly.  The W3130 printer connects to AC power (100V to 240V) with an included power adapter, and also features an included rechargeable Li-Ion battery.
Catalog No. 50-302-9579