Learn More
Medchemexpress LLC Sericoside | 55306-04-2 | 666.84 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN104965MG
Encompass_Preferred
Sericoside is a triterpenoid with anti-inflammatory activity, which can be isolated from Terminalia. It exhibits strong lipolytic activity and can also reduce skin wrinkles and improve skin texture.
  • Anti-inflammatory activity
  • Strong lipolytic activity
  • Reduces skin wrinkles
  • Ameliorates skin texture
  • Stimulates glycerol release into the cytosol from deposited triglyceride in fully differentiated 3T3-L1 cells
Catalog No. 50-004-16952