Learn More
Medchemexpress LLC Serine Hydrolase inhibitor-21 | 366448-34-2 | C18H12N2O2S | 5 MG

Supplier: Medchemexpress LLC HY1534335MG
Serine Hydrolase inhibitor-21 (compound 8) is a pyridine-based serine hydrolase inhibitor. It exhibits a Ki of 429 nM for BuChE and has potential for use in Alzheimer's disease research.
- High purity (99.71%)
- White to off-white solid
- Soluble in DMSO
- Molecular weight of 320.37
- Recommended storage at 4°C (solid) or -80°C/-20°C (in solvent)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.