Learn More
Abcam Serine Protease Assay Kit (SR101-Leu-CMK), 25 Tests
Supplier:  Abcam AB27077625TEST
Encompass_Preferred
These Serine Protease Assay Kits enable researchers to detect intracellular chymotrypsin-like serine protease activity in vitro without lysing the cell.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1981