Learn More
Chemglass Life Sciences Serological Pipette, 25mL, Color Coded Red, Individually Wrapped, Sterile
SDP
Supplier:  Chemglass Life Sciences CGN3481025
Encompass_Preferred
Serological pipettes are made of super clear clarity polystyrene (PS). Serological pipettes are designed to be used in both tissue culture and bacterial culture experiments, and other liquid handling applications.Edged color-coded band for identificationInclude negative graduations for extra volumeNon-pyrogenic, non-cytotoxic, non-hemolytic, DNase- and RNase-freePolyolefin fiber filter to reduce contaminationMade from high clarity polystyreneSterilized by E-beam
Catalog No. 50-189-7701