Learn More
Bellco Glass, Inc. Serological Shortie Pipet, 2x.1mL
SDP
Supplier:  Bellco Glass, Inc. 122402010
Encompass_Preferred
Bellco manufactures all precision pipettes from type 1 class a Borosilicate glass. All bellco pipettes feature tempered tips and permanent silk screened graduations to withstand repeated usage and alkali washings. Plain top promotes rapid, even flow (use rubber bulb to fill). Calibrated to tip. Tempered to deliver blow out. Case Qty: 18 TOLERANCE +-(ML):0.025OAL (MM):160COLOR CODE: Green Graduation: 0.1 MADE IN THE USA
Catalog No. 50-194-4609