Learn More
USA Scientific Inc 50 mL serological pipet, sterile
Supplier:  USA Scientific Inc 10571010
Encompass
50 mL serological pipet, standard tip orifice, 0.5 mL graduations, individually wrapped, sterile, 25/sleeve, 100/case
Catalog No. 50-754-1547