Learn More
Vee Gee Serological Pipet, 25 mL, 0.1 graduation interval
Supplier:  Vee Gee 2011A25/DEL
Encompass_Preferred
The Serological Pipet delivers and blows out. It meets ASTM E-1044 Style 1, Class A Specifications.: 25.0 mL • Tolerance: ± 0.05 mL • Graduation Interval: 0.1 mL • Approx. O.D. x Height: 13 x 455 mm • Color Code: White
Catalog No. NC1937452