Learn More
Thomas Scientific Mini Serological Pipettes, 5mL, Individually Wrapped
SDP
Supplier:  Thomas Scientific 1213N23
Encompass_Preferred
Argos brand mini disposable pipettes were developed for tissue culture applications or for use in confined spaces such as laminar hoods. These mini pipettes measure 9” and fit most serological pipettors. Smaller size reduces distance between sample and pipettor aiding in more ergonomic pipetting. Product Details: Polystyrene, gamma sterilized & pyrogen free Easy to read graduations from +1 - 2% Unique filter plug stays put and eliminates lint fibers Color coded rings for easy identification Fiber free paper & plastic wrap ensures quick peel apart opening Dispenser box allows for access of pipettes without opening top; color coded with universal stacking design
Catalog No. 50-146-0787