Learn More
Selleck Chemical LLC Serotonin
SDP
Supplier:  Selleck Chemical LLC S635125MG
Encompass_Preferred
Serotonin is a monoamine neurotransmitter in the Central Nervous System (CNS) and an endogenous 5-HT receptor agonist Serotonin is also a catechol O-methyltransferase (COMT) inhibitor
Catalog No. 50-000-04121