Learn More
Medchemexpress LLC Serotonin-d4 | 58264-95-2 | 99.6% | 1 MG
SDP
Supplier:  Medchemexpress LLC HYB1473AS1MG
Encompass_Preferred
Serotonin-d4 is a deuterium labeled Serotonin, functioning as a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. It also acts as a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. Stable heavy isotopes like deuterium are incorporated into drug molecules as tracers for quantitation during drug development, with potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes into drug molecules for quantitation
  • Deuteration can affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-004-11179