Learn More
Medchemexpress LLC Serotonin hydrochloride | 153-98-0 | 99.9% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB147350MG
Encompass_Preferred
Serotonin (5-Hydroxytryptamine) hydrochloride is an analytical standard and a monoamine neurotransmitter in the CNS. It acts as an endogenous 5-HT receptor agonist and a catechol O-methyltransferase (COMT) inhibitor with a Ki of 44 μM. This compound is used for research and analytical applications, including qualitative, quantitative, and methodological experiments in HPLC, GC, and MS. Its blood-brain barrier permeability is noted.
  • Used as an analytical standard
  • Functions as a monoamine neurotransmitter in the CNS
  • Acts as an endogenous 5-HT receptor agonist
  • Inhibits catechol O-methyltransferase (COMT)
  • Suitable for HPLC, GC, and MS research experiments
Catalog No. 50-004-11353