Learn More
Medchemexpress LLC Serotonin hydrochloride | 153-98-0 | 99.97% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB147310MM/1ML
Encompass_Preferred
Serotonin hydrochloride (5-Hydroxytryptamine hydrochloride) is a monoamine neurotransmitter in the CNS and an endogenous 5-HT receptor agonist. It possesses blood-brain barrier permeability and also acts as a catechol O-methyltransferase (COMT) inhibitor.
  • Functions as a monoamine neurotransmitter.
  • Acts as an endogenous 5-HT receptor agonist.
  • Exhibits blood-brain barrier permeability.
  • Inhibits catechol O-methyltransferase (COMT) with a Ki of 44 μM.
  • Decreases COMT reaction velocity at 100 μM concentration.
  • Offers high purity (99.97%).
  • Available as solid or solution in DMSO.
Catalog No. 50-004-11352