Learn More
Enzo Life Sciences Serotonin. hydrochloride (50mg). CAS: 153-98-0
SDP
Supplier:  Enzo Life Sciences ALX550328M050
Encompass_Preferred
Neurotransmitter. Endogenous 5-HT agonist produced by serotonergic neurons in the CNS and enterochromaffin cells in the gastrointestinal tract. Alternative name: 5-Hydroxytryptamine . HCl. Purity: ≥98% (HPLC). Solubility: Soluble in water, methanol or 100% ethanol. Long Term Storage: +4°C.
Catalog No. 50-200-9536