Learn More
Rocky Mountain Diagnostics Inc Serotonin High Sensitive ELISA Kit, competitive assay, 96-well format, Sensitivity 5 pg/ml, Sample Volume 1 μl, Storage 2-8°C for 6 months
SDP
Supplier:  Rocky Mountain Diagnostics Inc BA E5900R
Encompass
Serotonin High Sensitive ELISA Kit, competitive assay, 96-well format, Sensitivity 5 pg/ml, Sample Volume 1 μl, Storage 2-8°C for 6 months
Catalog No. NC2022260