Learn More
Medchemexpress LLC Serpin B5/Maspin Hu 100ug
SDP
Supplier:  Medchemexpress LLC HYP7385100UG
Encompass_Preferred
Serpin B5/Maspin Protein Human (375 a a) could inhibit invasion and motility of mammary tumors Maspin Protein Human is also found to be a potent angiogenesis inhibitor
Catalog No. 50-001-79008