Learn More
Medchemexpress LLC Serpin F1 Human HE 50ug
SDP
Supplier:  Medchemexpress LLC HYP70498A50UG
Encompass_Preferred
Recombinant human PEDF/Serpin F1 protein, expressed in HEK293 cells with an N-terminal His tag and supplied lyophilized for research use. The protein is active in cell-based adhesion assays and is provided with low endotoxin to support sensitive biological experiments.
  • Recombinant human PEDF/Serpin F1 expressed in HEK293 cells.
  • N-terminal His tag for detection and purification.
  • Lyophilized formulation for long-term storage.
  • Low endotoxin level (<1 EU/μg) suitable for cell assays.
  • Active in Saos-2 cell adhesion assay (ED50 0.2506 μg/mL).
  • Supplied as a 50 μg aliquot; recommended storage at -20°C.
Catalog No. 50-001-85502