Learn More
Apexbio Technology LLC Sertaconazole nitrate 99592-39-9 50mg
SDP
Supplier:  Apexbio Technology LLC B183150
Encompass_Preferred
Sertaconazole nitrate (CAS 99592-39-9) is a small-molecule inhibitor targeting lanosterol 14 -demethylase It is designed to inhibit fungal ergosterol biosynthesis thereby disrupting fungal cell membrane synthesis and integrity Sertaconazole nitrate exerts its biological activity primarily through interference with lanosterol 14 -demethylase and direct interaction with fungal cell membranes In in vitro studies sertaconazole nitrate demonstrates inhibitory activity with IC50 values generally ranging between 0 1 2 g/mL against various pathogenic fungal organisms Based on these pharmacological properties sertaconazole nitrate holds research potential in antifungal susceptibility testing and experimental studies of dermatophytes and superficial mucocutaneous fungal infections
Catalog No. 50-000-92266