Learn More
TARGETMOL CHEMICALS INC SERTINDOLE 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T585825MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Sertindole (Lu 23-174) is an atypical antipsychotic that binds to dopamine D2 receptors and the serotonin (5-HT) receptor subtypes 5-HT1D 5-HT2A and 5-HT2C (Kds = 2.7 20 0.14 and 6 nM respectively)purity: 99%
Catalog No. 50-257-9726