Learn More
Enzo Life Sciences Sertraline . HCl (10mg). CAS: 79559-97-0
SDP
Supplier:  Enzo Life Sciences BMLNS1150010
Encompass_Preferred
A selective serotonin transporter inhibitor. In clinical use as an antidepressant. Displays antiproliferative activity on human colorectal carcinoma cells. Alternative name: CP-51974. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (>25mg/ml), ethanol (10mg/ml) or water (3mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-1211