Learn More
Medchemexpress LLC Serum Albumin/ALB R 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP787371MG
Encompass_Preferred
ALB protein is a serum protein synthesized by the liver and plays a vital role in maintaining body fluid balance and transporting various molecules in the blood It is involved in the regulation of osmotic pressure and acts as a carrier for hormones fatty acids and drugs Serum Albumin/ALB Protein Rabbit (HEK293 His) is the recombinant Rabbit-derived Serum Albumin/ALB protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-94042