Learn More
Abcam Serum Antibody Purification Kit (Protein A), 3 Tests
Supplier:  Abcam AB1092093TEST
Encompass_Preferred
Serum Antibody Purification Kit (Protein A) (ab109209) is prepared by coupling highly purified protein A to agarose beads and can therefore be used to purify IgG fractions from both serum and ascites fluid.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-5211