Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Electron Microscopy Sciences Serum Bottles Clear Wheaton 400, 100 ml, 52 mm diameter, 95 mm height
SDP

Supplier:  Electron Microscopy Sciences 60306100

Encompass_Preferred

Manufactured from Wheaton borosilicate molded glass, meets USP Type I standards.
Volume: 100 ml
Diameter: 52 mm
Height: 95 mm
Material: Wheaton "400"
Cap: 13 x 20 mm

Catalog No. 50-189-8937


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.