Learn More
STEMCELL Technologies STEMdiff™ Myogenic Progenitor Supplement Kit, Size: 1 Kit
SDP
Supplier:  STEMCELL Technologies 1000151
Encompass
STEMdiff™ Myogenic Progenitor Supplement Kit comprises serum-free supplements for the differentiation of human embryonic stem (ES) cells and induced pluripotent stem (iPS) cells to myogenic progenitor cells. These cells, which are characterized by myogenic cell markers such as CD56 and CD82, can be culture-expanded for 5+ passages using MyoCult™-SF Expansion Supplement Kit (Human; Catalog #05980) and further differentiated to functional multinucleated MyHC+ myotubes at high efficiency using MyoCult™ Differentiation Kit (Human; Catalog #05965). These myotubes can be used in various downstream applications and analyses. This kit is compatible with human ES and iPS cells maintained in mTeSR™ 1 (Catalog #85850), mTeSR™ Plus (Catalog #100-0276), or TeSR™-E8™ (Catalog #05990).
Catalog No. 50-204-5482