Learn More
Crescent Chemical Co Inc SERVA TRIS-GLYCINE/SDS BUFFER
SDP
Supplier:  Crescent Chemical Co Inc SE42529.01
Encompass_Preferred
SERVA Tris-Glycine/SDS electrophoresis buffer (10x) Running buffer for SDS PAGE. Supplied as 10 x concentrate. Contains 250 mM Tris, 1.92 M glycine, 1 % SDS. Size - 1L Storage Conditions - +15 °C TO +30 °C Catalog Number - 42529.01
Catalog No. 50-248-593