Learn More
Crescent Chemical Co Inc SERVALYT™ 6-7
SDP
Supplier:  Crescent Chemical Co Inc SE42925.02
Encompass_Preferred
SERVALYT™ 6-7 40 % (w/v) solution in water SERVALYT™ Carrier Ampholytes (40 % w/v in water). Standard quality (analytical grade) for general use in IEF. Size - 25ML Storage Conditions - +2 °C TO +8 °C Catalog Number - 42925.02
Catalog No. 50-248-683