Learn More
Medchemexpress LLC Sesamolin | 526-07-8 | 370.35 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN080910MM/1ML
Encompass_Preferred
Sesamolin, isolated from Sesamum indicum, possesses antioxidative activity and inhibits lipid peroxidation. It also demonstrates a neuroprotective effect. Sesamolin potently inhibits MAPK cascades by preventing the phosphorylation of JNK, p38 MAPKs, and caspase-3, but not ERK-MAPK expression. It is orally active.
  • Antioxidative activity
  • Inhibits lipid peroxidation
  • Shows neuroprotection effect
  • Potently inhibits MAPK cascades by preventing phosphorylation of JNK, p38 MAPKs, and caspase-3
  • Orally active
Catalog No. 50-004-16686