Learn More
World Precision Instrument Set of 8 Calcium Buffers
SDP
Supplier:  World Precision Instrument CALBUF1
Encompass
A set of eight calcium buffers covering the range of concentration from 10-1 to 10-8 M Ca++. Each buffer contains 20 mL of solution and enough potassium chloride to set the ionic strength to 0.1 M.
Catalog No. 50-252-1210