Learn More
Quality Lab Accessories SET SCREW STYLE CANNULA STOPPR
Supplier:  Quality Lab Accessories CANSTPST
Encompass
Adjustable Cannula Stopper with set screw for Sotax (use with CAN013-ST,.125" dia.
Catalog No. NC2242655