Learn More
Medchemexpress LLC Setanaxib | 1218942-37-0 | 99.8% | 1 G
SDP
Supplier:  Medchemexpress LLC HY122981G
Encompass_Preferred
Setanaxib is a selective small-molecule inhibitor of NADPH oxidase isoforms NOX1 and NOX4 for research use. It is used to probe reactive oxygen species-mediated signaling and pathophysiology in studies of fibrosis, inflammation, and oxidative stress. Reported Ki values are 140 nM for NOX1 and 110 nM for NOX4, and the compound is commonly supplied as a 1 g research sample.
  • Selective NOX1/NOX4 inhibition with reported Ki values of 140 nM and 110 nM.
  • Suitable for biochemical and cell-based assays.
  • Used in research on oxidative stress, fibrosis, and inflammation.
  • High purity for reliable experimental results.
  • Provided as a 1 g research reagent.
Catalog No. 50-002-33749