Learn More
Compass Instruments SETAVAP4 AUTOMATIC ANALYZER
SDP
Supplier:  Compass Instruments SS806002
Encompass
SetaVap4 Automatic Vapor Pressure Analyzer, according to ASTM D5191, D5188, D6377, D6378, IP 394, EN 13016-1 & 3. Includes waste container, sample inlet tube, mains lead and operating manual.• Rapid automatic vapor pressure testing• 0 to 1000 kPa• Piston based design for triple and single expansion test methods• Peltier cells for heating and cooling• Integrated shaker for crude oil measurements• 8.4” touchscreen display• Integrated statistical quality control (SQS) software, compliant with ASTM D6299• Simple test procedure• Internal result storage with USB port for data download• Compact, rugged design ideal for portable field or laboratory use• DVPE, RVP, EPA and CARB calculations• For operation at 115V, 60 Hz.
Catalog No. NC3353236
Only null left
Add to Cart
Request a Quote