Learn More
Cayman Chemical Sethyl IsothIoureahydrobro
SDP
Supplier:  Cayman Chemical 81275500 MG
Encompass_Preferred
SEIT is a potent inhibitor of NOS in vitro but lacks good in vivo efficacy due to poor cellular penetration. The Ki values are 19, 39, and 29 nM using purified human iNOS, eNOS, and nNOS, respectively.{1292,1351,2259}
Catalog No. 50-289-5542